Learn More
Aviva Systems Biology Corp REN Antibody - C-terminal region (ARP41409_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41409_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against REN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human REN. Alternate names: FLJ10761, HNFJ2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-57838