Learn More
Biogenex Laboratories Renal Cell Carc : RCC-26 : 6mL
SDP
Catalog No. 501978861
Encompass_Preferred
RenalCell Carcinoma antibody recognizes a 200 kDa glycoprotein localized in the brush border of the proximal renal tubule. This antibody immunoreacts with approximately 90% of primary renal cell carcinomas and approximately 85% of metastatic renal cellcarcinomas. Other tumors that may react with this antibody are parathyroid adenoma, an occasional breast carcinoma. Nephroblastoma, oncocytoma, mesoblastic nephroma, transitional cell carcinoma, and angiomyolipoma are not labeled with this antibody.
Catalog No. 50-197-8861 Supplier Biogenex Laboratories Supplier No. AM5435M
RenalCell Carcinoma antibody recognizes a 200 kDa glycoprotein localized in the brush border of the proximal renal tubule. This antibody immunoreacts with approximately 90% of primary renal cell carcinomas and approximately 85% of metastatic renal cellcarcinomas. Other tumors that may react with this antibody are parathyroid adenoma, an occasional breast carcinoma. Nephroblastoma, oncocytoma, mesoblastic nephroma, transitional cell carcinoma, and angiomyolipoma are not labeled with this antibody.