Learn More
Diagnostic Biosystems RENAL CELL CARCINOMA Renal Cell Carcinoma (gp200)
Catalog No. 502407450
Encompass_Preferred
This antibody recognizes a 200 kD glycoprotein (gp200) localized in the brush border of the proximal renal tebules and the luminal surface of Bowman’s capsule. gp200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. Gp200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands and epididymis.
Catalog No. 50-240-7450 Supplier Diagnostic Biosystems Supplier No. PDM169
This antibody recognizes a 200 kD glycoprotein (gp200) localized in the brush border of the proximal renal tebules and the luminal surface of Bowman’s capsule. gp200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. Gp200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands and epididymis.