Learn More
Promocell GmbH PromoCell Renal Epithelial Cell Growth Medium 2 500 ml, ready-to-use, low-serum, with C-39606 attached to the flask
Catalog No. 50306248
Encompass
PromoCell Renal Epithelial Cell Growth Medium 2 is an optimized low-serum (5% V/V) medium that additionally contains Epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro making the Renal Epithelial Cell Growth Medium 2 the first choice for PromoCell Renal Epithelial Cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine and mouse renal epithelial cells.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 36 ng / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 4 pg / ml || Transferrin (recombinant human) | 5 μg / ml |
Catalog No. 50-306-248 Supplier Promocell GmbH Supplier No. C26030
PromoCell Renal Epithelial Cell Growth Medium 2 is an optimized low-serum (5% V/V) medium that additionally contains Epinephrine. The combination of low serum and selected growth factors ensures successful growth in vitro making the Renal Epithelial Cell Growth Medium 2 the first choice for PromoCell Renal Epithelial Cells.Although all PromoCell media are optimized for use with primary human cells, we have received feedback from customers that this particular medium can also be used for porcine and mouse renal epithelial cells.| Media supplement | Final supplement concentration|| Fetal Calf Serum | 0.05 ml / ml || Epidermal Growth Factor (recombinant human) | 10 ng / ml || Insulin (recombinant human) | 5 μg / ml || Hydrocortisone | 36 ng / ml || Epinephrine | 0.5 μg / ml || Triiodo-L-thyronine | 4 pg / ml || Transferrin (recombinant human) | 5 μg / ml |