Learn More
ATCC Renal Epithelial Cell Growth Kit
Catalog No. 502381931
Encompass
Renal Epithelial Cell Growth Kit contains components that when added to Renal Epithelial Cell Basal Medium (ATCC PCS-400-030) create a complete ATCC Primary Cell Solutions culture environment for renal epithelial cells derived from a normal human kidney (eg, Primary Renal Proximal Tubule Epithelial Cells, Normal, Human, ATCC PCS-400-010; Primary Renal Cortical Epithelial Cells, Normal, Human, ATCC PCS-400-011; Primary Renal Mixed Epithelial Cells, Normal, Human, ATCC PCS-400-012). The low serum (0.5% FBS) medium formulation is designed to support normal renal cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.
Catalog No. 50-238-1931 Supplier ATCC Supplier No. PCS400040
Renal Epithelial Cell Growth Kit contains components that when added to Renal Epithelial Cell Basal Medium (ATCC PCS-400-030) create a complete ATCC Primary Cell Solutions culture environment for renal epithelial cells derived from a normal human kidney (eg, Primary Renal Proximal Tubule Epithelial Cells, Normal, Human, ATCC PCS-400-010; Primary Renal Cortical Epithelial Cells, Normal, Human, ATCC PCS-400-011; Primary Renal Mixed Epithelial Cells, Normal, Human, ATCC PCS-400-012). The low serum (0.5% FBS) medium formulation is designed to support normal renal cell morphology as well as promote rapid growth and proliferation. No feeder layers, extracellular matrix proteins, or other substrates are required.