Learn More
Pyramex™ Rendezvous™ Safety Eyewear
Catalog No. 19155686 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Lens Coating:
Impact-resistant
Frame Color:
Black
Lens Tint:
Blue
Gray
Orange
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Coating Frame Color Lens Tint
19-155-686 Impact-resistant Black Gray
19-155-688 - Black Orange
19-155-690 Impact-resistant Black Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-155-686 Supplier Pyramex™ Supplier No. SB2820ST
Add to Cart
Add to Cart
This item is not returnable. View return policy
Fully adjustable spectacles
  • Temples adjust to four different lengths
  • Adjustable nose pad conforms to broad range of bridge sizes
  • Crafted from high grade polycarbonate, machined from virgin materials and formed to shape
  • Align lens angle to proper position for work application with ratcheting lens pitch adjustment
  • 9.5 base curve lens provides excellent side protection against wind and airborne particles
  • Provides 99.9% protection against harmful UV rays
  • Passes MIL-PRF 32432 High Velocity Impact Standards
  • Specifications
    Certifications/Compliance ANSI Z87.1
    Lens Coating Impact-resistant
    Lens Tint Gray
    Frame Color Black
    Weight (Metric) 28 g
    Lens Material Polycarbonate
    Frame Material Polycarbonate