Learn More
Alpco Ltd Renin ELISA, 96 Wells, 50 L;;
SDP
Supplier:  Alpco Ltd 20RENHUE01
Encompass_Preferred
The Renin ELISA is a sandwich enzyme-linked immunosorbent assay for measurement of active renin in human serum and plasma.For research use only. Not for use in diagnostic procedures.
  • Includes controls and standards
  • High analytical sensitivity of 0.80 pg/mL
  • Time saving incubating period just 3 hours and 15 minutes
  • Colorimetric detection
Research Use Only. Not for Use in Diagnostic Procedures.
Catalog No. 50-305-121