Learn More
Cayman Chemical Renin Fluorogenic Substrate (trifluoroacetate salt), Purity: ≥95%, 1 mg
SDP
Catalog No. 502890773
Encompass_Preferred
A probe for renin activity; consists of the normal peptide substrate for renin which has been linked to the fluorophore EDANS at one end and to a non-fluorescent quenching molecule (Dabcyl) at the other; can be used to quantify renin activity at ex/em = 340/485-510 nm, respectively
Catalog No. 50-289-0773 Supplier Cayman Chemical Supplier No. 100069061 MG
A probe for renin activity; consists of the normal peptide substrate for renin which has been linked to the fluorophore EDANS at one end and to a non-fluorescent quenching molecule (Dabcyl) at the other; can be used to quantify renin activity at ex/em = 340/485-510 nm, respectively