Learn More
CPC Scientific H-Arg-Arg-Pro-Phe-His-Sta-Ile-His-Lys-OH (trifluoroacetate salt) 5MG
SDP
Catalog No. 502110368
Encompass
This peptide is a potent human renin inhibitor. It is an analog of amino acid sequence around the cleavage site of the human angiotensinogen. The renin-angiotensin system plays an important role in blood pressure regulation. The inhibition of renin by this peptide lowers the blood pressure and therefore renin inhibitor III can contribute to the hypertension research.ONE-LETTER SEQUENCE: RRPFH-Sta-IHKMOLECULAR FORMULA: MOLECULAR WEIGHT:1247.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: Wood J. et al. Hypertension. 7, 797 (1985)
Catalog No. 50-211-0368 Supplier CPC Scientific Supplier No. RENI014A
This peptide is a potent human renin inhibitor. It is an analog of amino acid sequence around the cleavage site of the human angiotensinogen. The renin-angiotensin system plays an important role in blood pressure regulation. The inhibition of renin by this peptide lowers the blood pressure and therefore renin inhibitor III can contribute to the hypertension research.ONE-LETTER SEQUENCE: RRPFH-Sta-IHKMOLECULAR FORMULA: MOLECULAR WEIGHT:1247.6STORAGE CONDITIONS: -20 5CRESEARCH AREA: CardiovascularREFERENCES: Wood J. et al. Hypertension. 7, 797 (1985)