Learn More
CPC Scientific H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Leu-Tyr-Tyr-Ser-OH (trifluoroacetate salt) 1MG
SDP
Catalog No. 502110362
Encompass
ONE-LETTER SEQUENCE: DRVYIHPFHLLYYSMOLECULAR FORMULA: C89H123N21O21MOLECULAR WEIGHT:1823.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [110200-37-8], SYNONYMS: Preangiotensinogen (1-14) (rat), Angiotensinogen (1-14) (rat), Tetradecapeptide Renin Substrate (rat)RESEARCH AREA: CardiovascularREFERENCES: J. Bouhnik et al., Biochemistry, 20, 7010 (1981)
Catalog No. 50-211-0362 Supplier CPC Scientific Supplier No. RENI006A
ONE-LETTER SEQUENCE: DRVYIHPFHLLYYSMOLECULAR FORMULA: C89H123N21O21MOLECULAR WEIGHT:1823.1STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [110200-37-8], SYNONYMS: Preangiotensinogen (1-14) (rat), Angiotensinogen (1-14) (rat), Tetradecapeptide Renin Substrate (rat)RESEARCH AREA: CardiovascularREFERENCES: J. Bouhnik et al., Biochemistry, 20, 7010 (1981)