Learn More
ACROBiosystems Recombinant Protein;Mouse RENIN Protein;HEK293;100UG;REN,RENIN,REN,HNFJ2,FLJ10761,angiotensinogenase
Catalog No. 501657784
Encompass_Preferred
Recombinant Protein;Mouse RENIN Protein;100UG;Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Arg 402 (Accession # NP_112469.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7784 Supplier ACROBiosystems Supplier No. RENM5222100UG
Recombinant Protein;Mouse RENIN Protein;100UG;Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Arg 402 (Accession # NP_112469.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.