Learn More
ACROBiosystems Recombinant Protein;Human RENIN Protein;HEK293;1MG;REN,RENIN,REN,HNFJ2,FLJ10761,angiotensinogenase
Catalog No. 501657737
Encompass_Preferred
Recombinant Protein;Human RENIN Protein;1MG;Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Arg 406 (Accession # AAH33474).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7737 Supplier ACROBiosystems Supplier No. RENH52211MG
Recombinant Protein;Human RENIN Protein;1MG;Human RENIN, His Tag (REN-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 24 - Arg 406 (Accession # AAH33474).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 43.2 kDa. The protein migrates as 45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.