Learn More
MSC Renishaw CMM Ball Stylus, 3 mm Ruby, Stainless Steel, M2
Catalog No. 5000534212
Encompass
Straight styli are designed to inspect simple features where direct, unobstructed contact with a measured surface is possible. This stylus features a stainless steel stem for excellent rigidity, ideal for applications where weight is not critical. The ruby tip, an industry standard, is one of the hardest materials available and suitable for most applications; however, ruby tips are not recommended for scanning aluminum parts due to adhesive pickup.
  • Inspects simple features with direct, unobstructed contact.
  • Stainless steel stem provides excellent rigidity.
  • Ruby tip is an industry standard for hardness, suitable for most applications.
  • Not recommended for scanning aluminum parts due to adhesive pickup.
  • REVO recommended and compatible with RSH175, RSH250, RHS350, and RSH450 modules.
Catalog No. 50-005-34212 Supplier MSC Supplier No. 00551705
Straight styli are designed to inspect simple features where direct, unobstructed contact with a measured surface is possible. This stylus features a stainless steel stem for excellent rigidity, ideal for applications where weight is not critical. The ruby tip, an industry standard, is one of the hardest materials available and suitable for most applications; however, ruby tips are not recommended for scanning aluminum parts due to adhesive pickup.
  • Inspects simple features with direct, unobstructed contact.
  • Stainless steel stem provides excellent rigidity.
  • Ruby tip is an industry standard for hardness, suitable for most applications.
  • Not recommended for scanning aluminum parts due to adhesive pickup.
  • REVO recommended and compatible with RSH175, RSH250, RHS350, and RSH450 modules.