Learn More
MSC Renishaw CMM Ball Stylus, 6 mm Ball Diameter, 150 mm Length, M4 Thread
Catalog No. 5000530090
Encompass
This CMM Ball Stylus is designed for use with Machine Tool Probes. It features a 6 mm ball diameter, a 150 mm overall length, and an M4 thread. The stylus includes a ruby point and a carbon fiber stem, ensuring precise and reliable measurements.
  • Ruby point.
  • Carbon fiber stem.
  • M4 thread.
  • 6 mm ball diameter.
  • 150 mm overall length.
  • Suitable for machine tool probes.
Catalog No. 50-005-30090 Supplier MSC Supplier No. 59330381
This CMM Ball Stylus is designed for use with Machine Tool Probes. It features a 6 mm ball diameter, a 150 mm overall length, and an M4 thread. The stylus includes a ruby point and a carbon fiber stem, ensuring precise and reliable measurements.
  • Ruby point.
  • Carbon fiber stem.
  • M4 thread.
  • 6 mm ball diameter.
  • 150 mm overall length.
  • Suitable for machine tool probes.