Learn More
MSC Renishaw CMM Ball Stylus, M4, 6 mm Ruby Ball, 100 mm Length, Carbon Fiber
Catalog No. 5000537096
Encompass
This precision ball stylus is designed for high-accuracy probing applications on coordinate measuring machines (CMMs). Its 6 mm ruby ball offers excellent hardness and wear resistance for reliable contact and consistent measurement. The carbon fiber stem provides an exceptional stiffness-to-weight ratio, low mass, and reduced deflection, maintaining accuracy over extended lengths. With an overall length of 100 mm and an effective working length of 88.5 mm, it ensures extended reach with rigidity and measurement stability for precision metrology.
  • M4 thread type
  • 6 mm ruby ball for excellent hardness and wear resistance
  • Carbon fiber stem for exceptional stiffness-to-weight ratio, low mass, and reduced deflection
  • Extended reach while maintaining rigidity and measurement stability
  • Overall length of 100 mm
  • Effective working length of 88.5 mm
Catalog No. 50-005-37096 Supplier MSC Supplier No. 59330399
This precision ball stylus is designed for high-accuracy probing applications on coordinate measuring machines (CMMs). Its 6 mm ruby ball offers excellent hardness and wear resistance for reliable contact and consistent measurement. The carbon fiber stem provides an exceptional stiffness-to-weight ratio, low mass, and reduced deflection, maintaining accuracy over extended lengths. With an overall length of 100 mm and an effective working length of 88.5 mm, it ensures extended reach with rigidity and measurement stability for precision metrology.
  • M4 thread type
  • 6 mm ruby ball for excellent hardness and wear resistance
  • Carbon fiber stem for exceptional stiffness-to-weight ratio, low mass, and reduced deflection
  • Extended reach while maintaining rigidity and measurement stability
  • Overall length of 100 mm
  • Effective working length of 88.5 mm