Learn More
MSC Renishaw CMM Stylus Adapter, M2 Female, M5 Male, Stainless Steel, 10 mm OAL
Catalog No. 5000533308
Encompass
This CMM stylus adapter facilitates the connection of different CMM components, allowing a stylus with an M2 male thread to connect to a probe or extension with an M5 female thread, or vice-versa. This flexibility expands measurement configurations without requiring new styli or probes for various thread sizes.
  • Thread adapter type
  • M2 female thread size
  • M5 male thread size
  • Made from stainless steel
  • 10 mm overall length
  • Manufactured in Germany
Catalog No. 50-005-33308 Supplier MSC Supplier No. 00553271
This CMM stylus adapter facilitates the connection of different CMM components, allowing a stylus with an M2 male thread to connect to a probe or extension with an M5 female thread, or vice-versa. This flexibility expands measurement configurations without requiring new styli or probes for various thread sizes.
  • Thread adapter type
  • M2 female thread size
  • M5 male thread size
  • Made from stainless steel
  • 10 mm overall length
  • Manufactured in Germany