Learn More
MSC Renishaw CMM Stylus Extension, Ceramic, M2 Female M2 Male, 50 mm
Catalog No. 5000529757
Encompass
This CMM stylus extension is designed for precise contact with workpiece surfaces. Featuring an M2 female to M2 male thread, this ceramic extension has an overall length of 50 mm and a stem diameter of 3.00 mm, ensuring durability and accuracy for coordinate measuring machine applications.
  • Made from durable ceramic material
  • Features an M2 female to M2 male thread configuration
  • Measures 50 mm in overall length
  • Has a stem diameter of 3.00 mm
  • Ideal for coordinate measuring machine applications
Catalog No. 50-005-29757 Supplier MSC Supplier No. 00552612
This CMM stylus extension is designed for precise contact with workpiece surfaces. Featuring an M2 female to M2 male thread, this ceramic extension has an overall length of 50 mm and a stem diameter of 3.00 mm, ensuring durability and accuracy for coordinate measuring machine applications.
  • Made from durable ceramic material
  • Features an M2 female to M2 male thread configuration
  • Measures 50 mm in overall length
  • Has a stem diameter of 3.00 mm
  • Ideal for coordinate measuring machine applications