Learn More
MSC Renishaw CMM Taper Shank, 35.3 mm, CAT 50
Catalog No. 5000528307
Encompass
This CMM Taper Shank is designed for use with OMP40, OMP400, and RMP40 systems. It features a 35.3 mm size and CAT 50 taper, ensuring compatibility and secure mounting for Coordinate Measuring Machine applications.
  • Designed for OMP40, OMP400, and RMP40 systems.
  • Features a CAT 50 taper size.
  • Manufactured in the United States.
Catalog No. 50-005-28307 Supplier MSC Supplier No. 31241052
This CMM Taper Shank is designed for use with OMP40, OMP400, and RMP40 systems. It features a 35.3 mm size and CAT 50 taper, ensuring compatibility and secure mounting for Coordinate Measuring Machine applications.
  • Designed for OMP40, OMP400, and RMP40 systems.
  • Features a CAT 50 taper size.
  • Manufactured in the United States.