Learn More
MSC Renishaw Pointer Stylus, M2, Tungsten Carbide Tip, 30° Angle, 10 mm
Catalog No. 5000534297
Encompass
Pointer styli are designed to inspect threaded forms, specific points, and scribed lines. This stylus features a tungsten carbide tip, making it ideal for applications demanding a hardwearing interface with an inspected item.
  • M2 threaded for secure attachment
  • Tungsten carbide tip material for durability
  • Stainless steel stem material
  • 30° angle for specific inspection tasks
  • 10 mm overall length
  • Pointer diameter of 1.4 mm
  • Spherical end feature with 0.1 mm radius
Catalog No. 50-005-34297 Supplier MSC Supplier No. 00552166
Pointer styli are designed to inspect threaded forms, specific points, and scribed lines. This stylus features a tungsten carbide tip, making it ideal for applications demanding a hardwearing interface with an inspected item.
  • M2 threaded for secure attachment
  • Tungsten carbide tip material for durability
  • Stainless steel stem material
  • 30° angle for specific inspection tasks
  • 10 mm overall length
  • Pointer diameter of 1.4 mm
  • Spherical end feature with 0.1 mm radius