Learn More
MSC Renishaw Stylus Extension, M4 Thread, Ceramic, 30 mm
Catalog No. 5000534142
Encompass
This ceramic stylus extension for Coordinate Measuring Machines (CMM) provides additional reach for probing setups, allowing access to deep or complex features. Its design ensures alignment and stability, crucial for high accuracy measurements in varying conditions due to ceramic's high stiffness and low thermal expansion.
  • Provides additional reach for CMM applications.
  • Enables access to deep or complex features.
  • Ensures alignment and stability during probing.
  • Ceramic material offers high stiffness and low thermal expansion.
  • Excellent stability for high measurement accuracy.
  • 7.4 mm stem diameter enhances stiffness and reduces deflection.
  • Built-in crash protection: ceramic stem shatters instead of bending, minimizing damage.
Catalog No. 50-005-34142 Supplier MSC Supplier No. 00551986
This ceramic stylus extension for Coordinate Measuring Machines (CMM) provides additional reach for probing setups, allowing access to deep or complex features. Its design ensures alignment and stability, crucial for high accuracy measurements in varying conditions due to ceramic's high stiffness and low thermal expansion.
  • Provides additional reach for CMM applications.
  • Enables access to deep or complex features.
  • Ensures alignment and stability during probing.
  • Ceramic material offers high stiffness and low thermal expansion.
  • Excellent stability for high measurement accuracy.
  • 7.4 mm stem diameter enhances stiffness and reduces deflection.
  • Built-in crash protection: ceramic stem shatters instead of bending, minimizing damage.