Learn More
MSC Renishaw TP20 Probe Module, Low Force, 38 mm OAL, M8 Thread
Catalog No. 5000516735
Encompass
The TP20 is a compact, module-changing touch-trigger probe featuring a two-piece design with a probe body and detachable stylus module. This enables diverse stylus configurations and extensions for accessing complex part features. Modules can be changed manually or automatically without requalification. This low force module, identified by a green cap, is ideal for applications needing minimal trigger force for workpiece deflection measurement.
  • Compact module changing touch-trigger probe
  • Two-piece design with detachable stylus module(s)
  • Allows use of a range of stylus configurations and extensions
  • Modules can be changed manually or automatically
  • No requalification of stylus tip needed after module change
  • Identified by a green cap
  • Designed for applications requiring low trigger force to measure workpiece deflection
  • Supports steel and carbide styli up to 30 mm long
Catalog No. 50-005-16735 Supplier MSC Supplier No. 09285206
The TP20 is a compact, module-changing touch-trigger probe featuring a two-piece design with a probe body and detachable stylus module. This enables diverse stylus configurations and extensions for accessing complex part features. Modules can be changed manually or automatically without requalification. This low force module, identified by a green cap, is ideal for applications needing minimal trigger force for workpiece deflection measurement.
  • Compact module changing touch-trigger probe
  • Two-piece design with detachable stylus module(s)
  • Allows use of a range of stylus configurations and extensions
  • Modules can be changed manually or automatically
  • No requalification of stylus tip needed after module change
  • Identified by a green cap
  • Designed for applications requiring low trigger force to measure workpiece deflection
  • Supports steel and carbide styli up to 30 mm long