Learn More
Crescent Chemical Co Inc RENLAM M-2
SDP
Supplier:  Crescent Chemical Co Inc SE13825.02
Encompass_Preferred
Renlam M-1 Solvent-free, modified bisphenol A epoxy resin. Epoxy equivalent weight 232-250. Yields 3-dimensional crosslinking blocks.Non-toxic substitute for ARALDITE CY 212, which contained dibutyl phthalate. Renlam M-1 has identical properties as ARALDI
Catalog No. 50-247-307