Learn More
Biocare Medical, LLC Renoir Red Diluent - 100 ml
SDP
Catalog No. 5082983
Encompass_Preferred
Renoir Red Diluent is specifically formulated to enhance certain types of antibodies and may increase titers up to 2 to 4 times. The diluent is a colored-coded red for identification. It is specially designed to work with Tris-based antibodies, rinsing buffers and all detection systems. It can also be used on an automated staining system.
Catalog No. 50-829-83 Supplier Biocare Medical, LLC Supplier No. PD904L
Renoir Red Diluent is specifically formulated to enhance certain types of antibodies and may increase titers up to 2 to 4 times. The diluent is a colored-coded red for identification. It is specially designed to work with Tris-based antibodies, rinsing buffers and all detection systems. It can also be used on an automated staining system.