Learn More
Innovative Research Inc RenProren 2x Dplt Hmn Pla 10ml
Catalog No. 502744040
Encompass_Preferred
Renin/Prorenin (REN/PREN) Double Depleted Human Plasma 10ml - Renin/Prorenin (REN/PREN) Double Depleted Human Plasma from Innovative Research is prepared from high-quality, citrated human plasma using immobilized anti-human Renin IgG.
Catalog No. 50-274-4040 Supplier Innovative Research Inc Supplier No. IRENPRENDP10ML
Renin/Prorenin (REN/PREN) Double Depleted Human Plasma 10ml - Renin/Prorenin (REN/PREN) Double Depleted Human Plasma from Innovative Research is prepared from high-quality, citrated human plasma using immobilized anti-human Renin IgG.