Learn More
Medchemexpress LLC HY-106446 10mg Medchemexpress, Rentiapril CAS:80830-42-8 Purity:>98%
SDP
Catalog No. 501968346
Encompass_Preferred
Medchemexpress, HY-106446 10mg Rentiapril CAS:80830-42-8 Rentiapril is an orally active angiotensin converting enzyme (ACE) inhibitor with antihypertensive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8346 Supplier Medchemexpress LLC Supplier No. HY10644610MG
Medchemexpress, HY-106446 10mg Rentiapril CAS:80830-42-8 Rentiapril is an orally active angiotensin converting enzyme (ACE) inhibitor with antihypertensive activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.