Learn More
Medchemexpress LLC Rentosertib | 2828567-39-9 | 99.96% | 487.57 | 250 MG
SDP
Catalog No. 5000394309
Encompass_Preferred
Rentosertib (INS018 055) (compound 112) is an orally active TNIK and MAP4K4 inhibitor with IC50 values of 12-120 nM for both targets. It is for research use only.
  • Orally active TNIK and MAP4K4 inhibitor.
  • Reduces TGF-β-induced α-SMA protein expression with an IC50 of 27.14 nM in MRC-5 cells.
  • Decreases N-cadherin and p-SMAD2/SMAD3 expression, increases E-cadherin expression, and inhibits TGF-β-induced epithelial-to-mesenchymal transition (EMT) and fibroblast-to-myofibroblast transition (FMT) in A549 cells.
  • Improves lung function and ameliorates pulmonary fibrosis in Bleomycin-administrated mouse models (3-30 mg/kg, po, twice daily for 2 weeks).
  • Reduces lymphocytes in bronchoalveolar lavage fluid (BALF), inhibits pro-inflammatory cytokines, and exhibits anti-inflammatory effects in LPS-induced mouse acute lung injury models (3-30 mg/kg, po, single dose).
Catalog No. 50-003-94309 Supplier Medchemexpress LLC Supplier No. 250MGHY160623
Add to Cart
Add to Cart
Rentosertib (INS018 055) (compound 112) is an orally active TNIK and MAP4K4 inhibitor with IC50 values of 12-120 nM for both targets. It is for research use only.
  • Orally active TNIK and MAP4K4 inhibitor.
  • Reduces TGF-β-induced α-SMA protein expression with an IC50 of 27.14 nM in MRC-5 cells.
  • Decreases N-cadherin and p-SMAD2/SMAD3 expression, increases E-cadherin expression, and inhibits TGF-β-induced epithelial-to-mesenchymal transition (EMT) and fibroblast-to-myofibroblast transition (FMT) in A549 cells.
  • Improves lung function and ameliorates pulmonary fibrosis in Bleomycin-administrated mouse models (3-30 mg/kg, po, twice daily for 2 weeks).
  • Reduces lymphocytes in bronchoalveolar lavage fluid (BALF), inhibits pro-inflammatory cytokines, and exhibits anti-inflammatory effects in LPS-induced mouse acute lung injury models (3-30 mg/kg, po, single dose).