Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International RenuCycle Green Giant

Catalog No. 19365513 Shop All Hourglass International Inc Products
Change view
Click to view available options
Capacity (English):
400 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-365-513 400 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Avoid placing PPE contaminated with hazardous chemicals such as acids, toxins, or infectious agents.

Catalog No. 19-365-513 Supplier Hourglass International Supplier No. RC433
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Avoid placing PPE contaminated with hazardous chemicals such as acids, toxins, or infectious agents.

Large-scale PPE recycling container capable of holding up to 400 lbs.

The RenuCycle Green Giant from Hourglass International is designed for businesses requiring large-scale PPE waste management.

  • Capacity: Up to 400 lb.
  • Dimensions: 4 x 3.3 x 3 ft.
  • Sturdy build
  • Diverts substantial PPE waste from landfills, aiding in circular economy efforts
  • Compatibility with Various PPE, including gloves, masks, hoods, and boot covers
  • Easy handling

Specifications

Capacity (English) 400 lb.
Product Type Recycling and Sustainability

Supports large-scale eco-friendly waste solutions for significant environmental impact.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.