Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International RenuCycle PPE Pillar

Catalog No. 19365514 Shop All Hourglass International Inc Products
Change view
Click to view available options
Capacity (English):
40 lb.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English)
19-365-514 40 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Do not use the PPE Pillar for items contaminated with hazardous substances such as caustic, toxic, or infectious chemicals.

Catalog No. 19-365-514 Supplier Hourglass International Supplier No. RC113
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Do not use the PPE Pillar for items contaminated with hazardous substances such as caustic, toxic, or infectious chemicals.

Compact vertical recycling solution, capable of holding up to 40 lbs of PPE.

The PPE Pillar simplifies PPE collection with these features:

  • Durable construction
  • Contributes to landfill diversion and sustainable practices
  • Easy to set up and maintain
  • Compatibility: Accepts all types of PPE, including gloves, masks, and non-woven fabrics.
  • Capacity: Up to 40 lb.
  • Dimensions: 1 x 1 x 3 ft.

Specifications

Capacity (English) 40 lb.
Product Type Recycling and Sustainability

Helps your facility contribute to eco-friendly practices and reduce its carbon footprint.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.