Learn More
Strem, An Ascensus Company CAS# 18960-54-8. 25g. Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)cerium(IV), min. 97% (99.9%-Ce) (REO) [Ce(TMHD)4]. MFCD00145378
SDP
Supplier:  Strem, An Ascensus Company 58500025G
Encompass
CAS# 18960-54-8. 25g. Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)cerium(IV), min. 97% (99.9%-Ce) (REO) [Ce(TMHD)4]. MFCD00145378. Molecular Weight: 873.20. Molecular Formula: Ce(C11H19O2)4. Color/form: red pwdr. Strem# 58-5000. http://www.strem.com/catalog/v/58-5000/
Catalog No. 50-204-5373