Learn More
Cayman Chemical Repaglinide, Purity: ≥98%, CAS: 135062-02-1, 100 mg
SDP
Catalog No. 502982368
Encompass_Preferred
An ATP-dependent potassium channel blocker (Kd = 0.42 nM for the sulphonylurea receptor SUR1 when co-expressed with Kir6.2); lowers blood glucose in fasted rats and dogs (ED50s = 10 and 28.3 μg/kg, respectively)
Catalog No. 50-298-2368 Supplier Cayman Chemical Supplier No. 19387100 MG
An ATP-dependent potassium channel blocker (Kd = 0.42 nM for the sulphonylurea receptor SUR1 when co-expressed with Kir6.2); lowers blood glucose in fasted rats and dogs (ED50s = 10 and 28.3 μg/kg, respectively)