Learn More
Apexbio Technology LLC Repaglinide 135062-02-1 100mg
SDP
Catalog No. 502240501
Encompass_Preferred
Repaglinide (CAS 135062-02-1) is a small-molecule modulator that acts as a blocker of ATP-sensitive potassium (K sub ATP /sub ) channels specifically binding with high affinity to the sulfonylurea receptor 1 (SUR1) subunit when co-expressed with K sub ir /sub 6 2 (K sub d /sub 0 42 nM) Through inhibition of K sub ATP /sub channel activity repaglinide promotes membrane depolarization and subsequent insulin secretion from pancreatic -cells In vivo studies demonstrate its glucose-lowering (hypoglycaemic) effects supporting its utility in antidiabetic research and in mechanistic investigations of pancreatic K sub ATP /sub channel regulation
This item is not returnable. View return policy
Catalog No. 50-224-0501 Supplier Apexbio Technology LLC Supplier No. A5316100
Repaglinide (CAS 135062-02-1) is a small-molecule modulator that acts as a blocker of ATP-sensitive potassium (K sub ATP /sub ) channels specifically binding with high affinity to the sulfonylurea receptor 1 (SUR1) subunit when co-expressed with K sub ir /sub 6 2 (K sub d /sub 0 42 nM) Through inhibition of K sub ATP /sub channel activity repaglinide promotes membrane depolarization and subsequent insulin secretion from pancreatic -cells In vivo studies demonstrate its glucose-lowering (hypoglycaemic) effects supporting its utility in antidiabetic research and in mechanistic investigations of pancreatic K sub ATP /sub channel regulation