Learn More
TARGETMOL CHEMICALS INC Repaglinide 200MG
SDP
Catalog No. 502880698
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Repaglinide (AG-EE 623ZW) is a benzoic acid derivative that stimulates insulin secretion from the pancreas and is used in the therapy of type 2 diabetes. Repaglinide has been linked to rare instances of clinically apparent acute liver injury. Purity 99.66%
Catalog No. 50-288-0698 Supplier TARGETMOL CHEMICALS INC Supplier No. T1088200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Repaglinide (AG-EE 623ZW) is a benzoic acid derivative that stimulates insulin secretion from the pancreas and is used in the therapy of type 2 diabetes. Repaglinide has been linked to rare instances of clinically apparent acute liver injury. Purity 99.66%