Learn More
AdipoGen Repaglinide (250 mg)
SDP
Supplier:  AdipoGen CDXR0122M250
Encompass_Preferred
Chemical. CAS: 135062-02-1. Formula: C27H36N2O4. MW: 452.59. Repaglinide is a metaglitinide antidiabetic agent. It is a potent short-acting insulin secretagogue that acts by closing ATP-sensitive potassium (KATP) channels in the plasma membrane of the pancreatic beta cell. It represents a new class of insulin secretagogues, structurally unrelated to sulphonylureas, which were developed for the treatment of type 2 diabetes. Repaglinide shows also antioxidant and anti-inflammatory properties.
Catalog No. 50-205-4631