Learn More
Medchemexpress LLC Repaglinide-d5 | 1217709-85-7 | C27H31D5N2O4 | 1 MG
SDP
Catalog No. 5000372999
Encompass_Preferred
Repaglinide-d5 is deuterium labeled Repaglinide, an insulin secretagogue for the treatment of type-2 diabetes mellitus. It reduces postprandial glucose levels by enhancing the early phase of insulin secretion and increasing the total amount of insulin secreted.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-72999 Supplier Medchemexpress LLC Supplier No. HY15209S1MG
Repaglinide-d5 is deuterium labeled Repaglinide, an insulin secretagogue for the treatment of type-2 diabetes mellitus. It reduces postprandial glucose levels by enhancing the early phase of insulin secretion and increasing the total amount of insulin secreted.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS