Learn More
Medchemexpress LLC Repaglinide-d5 | 1217709-85-7 | C27H31D5N2O4 | 5 MG
SDP
Catalog No. 5000373619
Encompass_Preferred
Repaglinide-d5 is deuterium labeled Repaglinide, an insulin secretagogue used for treating type-2 diabetes mellitus. Repaglinide reduces postprandial glucose levels by enhancing the early phase of insulin secretion and increasing the total amount of insulin secreted.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-73619 Supplier Medchemexpress LLC Supplier No. HY15209S5MG
Repaglinide-d5 is deuterium labeled Repaglinide, an insulin secretagogue used for treating type-2 diabetes mellitus. Repaglinide reduces postprandial glucose levels by enhancing the early phase of insulin secretion and increasing the total amount of insulin secreted.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS