Learn More
Selleck Chemical LLC Repaglinide 50mg 135062-02-1 AG-EE 623 ZW
SDP
Catalog No. 508812
Encompass_Preferred
Repaglinide (AG-EE 623 ZW) is a potassium channel blocker, which lowers blood glucose by stimulating the release of insulin from the pancreas, used the treatment of type II diabetes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-881-2 Supplier Selleck Chemical LLC Supplier No. S142650MG
Repaglinide (AG-EE 623 ZW) is a potassium channel blocker, which lowers blood glucose by stimulating the release of insulin from the pancreas, used the treatment of type II diabetes. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.