Learn More
Fine Science Tools Repair Kit for Fine Forceps, 4 Piece Kit
SDP
Catalog No. NC9972021
Encompass
Product: Repair Kit for Dumont Forceps/ Item: 4 Piece Kit/ Feature: Sharpening Stone, O-Rings, Pliers & Instrument Oil Unit: Each
Catalog No. NC9972021 Supplier Fine Science Tools Supplier No. 2900000
Product: Repair Kit for Dumont Forceps/ Item: 4 Piece Kit/ Feature: Sharpening Stone, O-Rings, Pliers & Instrument Oil Unit: Each