Learn More
Thermo Scientific™ Repair Kits for Thermo Scientific™ Plunger-in-Needle Syringes
Catalog No. 365RK321 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
PIN Syringe 365A0331
PIN Syringe 365B0511
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
36-5RK-321 PIN Syringe 365A0331
36-5RK-311 PIN Syringe 365B0511
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 36-5RK-321 Supplier Thermo Scientific™ Supplier No. 365RK321
Extend the life and performance, and reduce the cost per use of your syringes with the Thermo Scientific™ Repair Kit for Thermo Scientific Plunger-in-Needle Syringes.
Thermo Scientific™ Repair Kits for Thermo Scientific Plunger-in-Needle Syringes extend the life and performance and reduce cost per use of syringes.
  • Repair kits for plunger in needle syringes contain both the plunger and needle, which must be changed together
Includes: Plunger, 1; Needle, 1
TRUSTED_SUSTAINABILITY
Specifications
Description For PIN syringe 365A0331
Needle Gauge 23 G
Length (English) Needle 1.9685 in.
Product Type Syringe Repair Kit
Volume (Metric) 0.5 μL
Length (Metric) Needle 50 mm
For Use With (Equipment) PIN Syringe 365A0331
Connection Type Plunger-In