Learn More
Apexbio Technology LLC Reparixin 266359-83-5 10mM (in 1mL DMSO)
SDP
Catalog No. 5000064310
Encompass_Preferred
Reparixin (CAS 266359-83-5) is a noncompetitive allosteric inhibitor of chemokine receptors CXCR1 and CXCR2 which are seven-transmembrane G-protein coupled receptors involved notably in acute inflammatory responses Reparixin specifically disrupts CXCR1/2-mediated neutrophil recruitment and migration without affecting other receptor pathways In murine ischemia-reperfusion injury models Reparixin administration significantly inhibits neutrophil influx and associated inflammation-related tissue damage Thus Reparixin serves as a valuable research tool for investigating CXCR-dependent inflammatory mechanisms and evaluating therapeutic potentials in conditions such as acute lung injury and spinal cord damage
Catalog No. 50-000-64310 Supplier Apexbio Technology LLC Supplier No. A37525.1
Reparixin (CAS 266359-83-5) is a noncompetitive allosteric inhibitor of chemokine receptors CXCR1 and CXCR2 which are seven-transmembrane G-protein coupled receptors involved notably in acute inflammatory responses Reparixin specifically disrupts CXCR1/2-mediated neutrophil recruitment and migration without affecting other receptor pathways In murine ischemia-reperfusion injury models Reparixin administration significantly inhibits neutrophil influx and associated inflammation-related tissue damage Thus Reparixin serves as a valuable research tool for investigating CXCR-dependent inflammatory mechanisms and evaluating therapeutic potentials in conditions such as acute lung injury and spinal cord damage