Learn More
Apexbio Technology LLC Reparixin, 200mg. Cas: 266359-83-5 MFCD: MFCD18633292. Inhibitor of CXCL8 receptor and CXCR1/CXCR2 activation
SDP
Supplier:  Apexbio Technology LLC A3752200
Encompass_Preferred
Reparixin is a non-competitive allosteric inhibitor of CXCR1/2.CXCR is a 7-transMEM G protein-coupled receptor. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4835