Learn More
Medchemexpress LLC Reparixin | 266359-83-5 | C14H21NO3S | 2 MG
SDP
Catalog No. 5000372952
Encompass_Preferred
Reparixin is a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation. It effectively inhibits CXCL8-induced biological activities on human PMNs, showing significant selectivity for CXCR1. This compound has been demonstrated to mitigate inflammatory responses across various injury models.
  • Non-competitive allosteric inhibitor of chemokine receptors CXCR1 and CXCR2
  • Potent functional inhibitor of CXCL8-induced biological activities
  • Attenuates inflammatory responses in various injury models
  • Decreases systolic blood pressure and increases blood flow in spontaneously hypertensive rats
  • Reduces IL-1β levels in the brain after middle cerebral artery occlusion/reperfusion in mice
Catalog No. 50-003-72952 Supplier Medchemexpress LLC Supplier No. HY152512MG
Reparixin is a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation. It effectively inhibits CXCL8-induced biological activities on human PMNs, showing significant selectivity for CXCR1. This compound has been demonstrated to mitigate inflammatory responses across various injury models.
  • Non-competitive allosteric inhibitor of chemokine receptors CXCR1 and CXCR2
  • Potent functional inhibitor of CXCL8-induced biological activities
  • Attenuates inflammatory responses in various injury models
  • Decreases systolic blood pressure and increases blood flow in spontaneously hypertensive rats
  • Reduces IL-1β levels in the brain after middle cerebral artery occlusion/reperfusion in mice