Learn More
TARGETMOL CHEMICALS INC REPARIXIN 5MG
SDP
Catalog No. 502576798
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reparixin (Repertaxin) is a potent inhibitor of both CXCL8 receptors CXCR1/2 it inhibits weakly CXCR2-mediated cell migration (IC50=100 nM) whereas it strongly blocks CXCR1-mediated chemotaxis (IC50=1 nM). purity: 99%
Catalog No. 50-257-6798 Supplier TARGETMOL CHEMICALS INC Supplier No. T41635MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Reparixin (Repertaxin) is a potent inhibitor of both CXCL8 receptors CXCR1/2 it inhibits weakly CXCR2-mediated cell migration (IC50=100 nM) whereas it strongly blocks CXCR1-mediated chemotaxis (IC50=1 nM). purity: 99%