Learn More
Medchemexpress LLC HY-15251 5mg Medchemexpress, Reparixin CAS:266359-83-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY152515MG
Encompass_Preferred
Medchemexpress, HY-15251 5mg Reparixin CAS:266359-83-5 Reparixin is a non-competitive allosteric inhibitor of the chemokine receptors CXCR1 and CXCR2 activation with IC50s of 1 and 100 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9108