Learn More
Selleck Chemical LLC Reparixin (Repertaxin) 2mg 266359-83-5
SDP
Supplier:  Selleck Chemical LLC S86402MG
Encompass_Preferred
Reparixin (Repertaxin) is a potent and specific inhibitor of CXCR1 with IC50 of 1 nM. Reparixin (Repertaxin) inhibits PMN migration induced by CXCL8 (IC50 = 1 nM) and rodent PMN chemotaxis induced by CXCL1 and CXCL2. Repertaxin inhibits the response of human PMN to CXCL1, which interacts with CXCR2 (IC50 = 400 nM). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8074