Learn More
USA Scientific Inc Repeater M4
Catalog No. 502745080
Encompass
Repeater M4 Starter Kit, 1-channel, Repeater M4, Combitips advanced Rack, Combitips advanced assortment pack, 1 uL-10 mL, holder for Carousel 2, Charger Carousel 2 or wall mounting
Catalog No. 50-274-5080 Supplier USA Scientific Inc Supplier No. 40492322
Repeater M4 Starter Kit, 1-channel, Repeater M4, Combitips advanced Rack, Combitips advanced assortment pack, 1 uL-10 mL, holder for Carousel 2, Charger Carousel 2 or wall mounting