Learn More
Pipette Supplies, Inc. 4 cell battery pack 1.2V Ni-Cd, 400mAh
SDP
Catalog No. 501317644
Encompass
4 cell battery pack 1.2V Ni-Cd, 400mAh. Battery must be charged prior to use. Eppendorf International Part# 4985801008, formerly EP 022466534.
Catalog No. 50-131-7644 Supplier Pipette Supplies, Inc. Supplier No. PS 022466534
Add to Cart
Add to Cart
4 cell battery pack 1.2V Ni-Cd, 400mAh. Battery must be charged prior to use. Eppendorf International Part# 4985801008, formerly EP 022466534.