Learn More
MilliporeSigma™ Supelco™ Repeating Dispenser
Catalog No. 111010454 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
For Use With (Equipment):
Hamilton PB600-1, 10μL to 2.5 mL Hamilton Syringes
Catalog No. For Use With (Equipment)
11-101-0454 Hamilton PB600-1, 10μL to 2.5 mL Hamilton Syringes
1 options
Catalog No. 11-101-0454 Supplier MilliporeSigma™ Supelco™ Supplier No. 20943
Add to Cart
Add to Cart
For use with Hamilton PB600-1, and all Hamilton 10 μL to 2.5 mL Syringes
  • Accurately divides samples
  • Discharges 1/50th of the syringe’s capacity with each push of the button
  • Fits all 10 μL to 2.5 mL Hamilton syringes
  • Syringes sold separately
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Accurately Dividing Samples
Product Type Repeating Dispenser
Quantity 1 Ea.
For Use With (Equipment) Hamilton PB600-1, 10μL to 2.5 mL Hamilton Syringes