Learn More
Electron Microscopy Sciences Repetitive Syringe Dispenser
SDP
Catalog No. 50949741
Encompass_Preferred
Syringe dispenser offers a fast and accurate way of dispensing multiple doses of a set-volume. The volume delivered is determined by the volume-selector-dial and the size of the syringe chosen. A chart comes with the unit to help you to choose the size of syringe, volume, and the number of dispenses. The set dispenses 21 different volumes up to 58 times without refilling, with an accuracy of +/- 1.5% or better depending on the size of the syringe. Technical Data tests are based on water at a temperature of 25 degree C. 0.6, 1.5, & 6.0ml syringes.
Catalog No. 50-949-741 Supplier Electron Microscopy Sciences Supplier No. 72480
Syringe dispenser offers a fast and accurate way of dispensing multiple doses of a set-volume. The volume delivered is determined by the volume-selector-dial and the size of the syringe chosen. A chart comes with the unit to help you to choose the size of syringe, volume, and the number of dispenses. The set dispenses 21 different volumes up to 58 times without refilling, with an accuracy of +/- 1.5% or better depending on the size of the syringe. Technical Data tests are based on water at a temperature of 25 degree C. 0.6, 1.5, & 6.0ml syringes.