Learn More
Aviva Systems Biology Corp REPIN1 Antibody - middle region (ARP31916_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31916_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against REPIN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human REPIN1. Alternate names: AP4, RIP60, ZNF464, Zfp464ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54370