Learn More
Medchemexpress LLC Repinotan hydrochloride | 144980-77-8 | 97.1% | 25 MG
SDP
Catalog No. 5000342738
Encompass_Preferred
Repinotan hydrochloride (BAY x 3702) is a potent, selective, brain-penetrant, and orally active 5-HT1A receptor agonist with Ki values of 0.19 nM (calf hippocampus), 0.25 nM (rat and human cortex), and 0.59 nM (rat hippocampus). It has a weak affinity for other related receptors and exhibits pronounced neuroprotective effects. It is intended for research use only.
  • Potent, selective, brain-penetrant, and orally active 5-HT1A receptor agonist.
  • Exhibits pronounced neuroprotective effects.
  • Weak affinity for other related receptors.
  • For research use only.
Catalog No. 50-003-42738 Supplier Medchemexpress LLC Supplier No. 25MGHY12959A
Repinotan hydrochloride (BAY x 3702) is a potent, selective, brain-penetrant, and orally active 5-HT1A receptor agonist with Ki values of 0.19 nM (calf hippocampus), 0.25 nM (rat and human cortex), and 0.59 nM (rat hippocampus). It has a weak affinity for other related receptors and exhibits pronounced neuroprotective effects. It is intended for research use only.
  • Potent, selective, brain-penetrant, and orally active 5-HT1A receptor agonist.
  • Exhibits pronounced neuroprotective effects.
  • Weak affinity for other related receptors.
  • For research use only.